Learn More
Medchemexpress LLC H-Phe 4-Ac -OH 100mg
SDP
Catalog No. 5000205067
Encompass_Preferred
H-Phe(4-Ac)-OH (L-4-Acetylphenylalanine) is a keto-amino acid that can be converted from -keto acids containing an acetyl group H-Phe(4-Ac)-OH can be added to the amber position to form mutant Z-domain proteins H-Phe(4-Ac)-OH is used as a functional amino acid in peptide modification to achieve chemical bonding between peptides and solid surfaces[1][2][3][4]
Catalog No. 50-002-05067 Supplier Medchemexpress LLC Supplier No. HYW102456100MG
H-Phe(4-Ac)-OH (L-4-Acetylphenylalanine) is a keto-amino acid that can be converted from -keto acids containing an acetyl group H-Phe(4-Ac)-OH can be added to the amber position to form mutant Z-domain proteins H-Phe(4-Ac)-OH is used as a functional amino acid in peptide modification to achieve chemical bonding between peptides and solid surfaces[1][2][3][4]