Learn More
Bachem H-Phe-Arg-OH
SDP
Supplier:  Bachem 4003975.0001
Encompass_Preferred
H-Phe-Arg-OH Trifluoroacetate, 1238-09-1, C15H23N5O3, Mr 321.38, 1g. Phe-Arg is cleaved from bradykinin by angiotensin-converting enzyme (ACE). See also Ser-Pro and RPPGF for futher cleavage products.
Catalog No. 50-382-194