Learn More
Medchemexpress LLC H-Phe-Asp-OH | 22828-05-3 | 99.08% | 280.28 | 10 MG
SDP
Catalog No. 5000398752
Encompass_Preferred
H-Phe-Asp-OH is a polypeptide that can be discovered through peptide screening, a research technique that primarily utilizes immunoassay to gather active peptides. This method is applicable for investigating protein interactions, conducting functional analyses, and performing epitope screening, particularly within the domain of agent research and development.
  • It is a polypeptide.
  • It can be found via peptide screening.
  • It serves as a research tool.
  • It is used for protein interaction studies.
  • It is utilized for functional analysis.
  • It is employed for epitope screening.
  • It is especially relevant in the field of agent research and development.
Catalog No. 50-003-98752 Supplier Medchemexpress LLC Supplier No. HYP447210MG
H-Phe-Asp-OH is a polypeptide that can be discovered through peptide screening, a research technique that primarily utilizes immunoassay to gather active peptides. This method is applicable for investigating protein interactions, conducting functional analyses, and performing epitope screening, particularly within the domain of agent research and development.
  • It is a polypeptide.
  • It can be found via peptide screening.
  • It serves as a research tool.
  • It is used for protein interaction studies.
  • It is utilized for functional analysis.
  • It is employed for epitope screening.
  • It is especially relevant in the field of agent research and development.