Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H-Phe-OMe.HCl, Advanced ChemTech

Catalog No. 01339644
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-339-644 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-339-644 Supplier Advanced Chem Tech Supplier No. IF7428100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7524-50-7 | C10H14ClNO2 | 215.68 g/mol

Chemical Identifiers

CAS 7524-50-7
Molecular Formula C10H14ClNO2
Molecular Weight (g/mol) 215.68
InChI Key SWVMLNPDTIFDDY-UHFFFAOYNA-N
IUPAC Name hydrogen methyl 2-amino-3-phenylpropanoate chloride
SMILES [H+].[Cl-].COC(=O)C(N)CC1=CC=CC=C1

Specifications

Color White to Off-White
Quantity 100 g
Grade Biochemical
Chemical Name or Material H-Phe-OMe.HCl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.