Learn More
Medchemexpress LLC H-Phg-OtBu.HCl | 161879-12-5 | 243.73 | 25 G
SDP
Catalog No. 5000449934
Encompass_Preferred
H-Phg-OtBu.HCl, also known as Benzeneacetic acid, α-amino-, 1,1-dimethylethyl ester, hydrochloride (1:1), (αS)-, is a high-purity glycine derivative. This white to off-white solid is suitable for various research applications and has been referenced in studies investigating the effects of amino acid derivatives on physical, mental, and physiological activities.
  • White to off-white solid
  • High purity (99.3% by HPLC)
  • Glycine derivative
  • Referenced in amino acid derivative research
Catalog No. 50-004-49934 Supplier Medchemexpress LLC Supplier No. 25GHYW006062
H-Phg-OtBu.HCl, also known as Benzeneacetic acid, α-amino-, 1,1-dimethylethyl ester, hydrochloride (1:1), (αS)-, is a high-purity glycine derivative. This white to off-white solid is suitable for various research applications and has been referenced in studies investigating the effects of amino acid derivatives on physical, mental, and physiological activities.
  • White to off-white solid
  • High purity (99.3% by HPLC)
  • Glycine derivative
  • Referenced in amino acid derivative research