Learn More
Chem-Impex International, Inc. L-Proline-2-chlorotrityl resin (0.3 - 0.8 meq/g, 100 - 200 mesh) | MFCD00801415 | 1G
Supplier:  Chem-Impex International, Inc. 034821G
Encompass_Preferred
L-Proline-2-chlorotrityl resin (0.3 - 0.8 meq/g, 100 - 200 mesh), MFCD00801415, 1G
Catalog No. 50-497-918