Learn More
Bachem H-Pro-Arg-OH
SDP
Supplier:  Bachem 4006116.0250
Encompass_Preferred
H-Pro-Arg-OH Acetate, 2418-74-8, C11H21N5O3, Mr 271.32, 250mg. PR inhibited angiotensin-1 converting enzyme (ACE), IC50 4.1 μM.
Catalog No. 50-382-209