Learn More
Medchemexpress LLC H-Pro-Hyp-OH | 18684-24-7 | MFCD00037349 | >99.0% | 228.25 g/mol | C10H16N2O4 | 100 MG
SDP
Catalog No. 5000282148
Encompass_Preferred
H-Pro-Hyp-OH (CAS 18684-24-7) is a collagen-derived dipeptide composed of proline and hydroxyproline (C10H16N2O4, 228.25 g/mol). It is supplied as a dry biochemical reagent for laboratory research, commonly used in studies of skin aging, fibroblast stimulation, and peptide-based assays. Consult the product datasheet and batch COA for purity and storage instructions.
  • Collagen-derived dipeptide for skin aging research
  • Well-defined small peptide suitable for biochemical assays
  • Available in multiple bench-friendly package sizes
  • Batch-specific COA available for purity verification
  • Compatible with standard peptide handling and storage protocols
Catalog No. 50-002-82148 Supplier Medchemexpress LLC Supplier No. HYW337672100MG
H-Pro-Hyp-OH (CAS 18684-24-7) is a collagen-derived dipeptide composed of proline and hydroxyproline (C10H16N2O4, 228.25 g/mol). It is supplied as a dry biochemical reagent for laboratory research, commonly used in studies of skin aging, fibroblast stimulation, and peptide-based assays. Consult the product datasheet and batch COA for purity and storage instructions.
  • Collagen-derived dipeptide for skin aging research
  • Well-defined small peptide suitable for biochemical assays
  • Available in multiple bench-friendly package sizes
  • Batch-specific COA available for purity verification
  • Compatible with standard peptide handling and storage protocols