Learn More
Bachem H-Pro-Hyp-OH
SDP
Catalog No. 50382217
Encompass_Preferred
H-Pro-Hyp-OH, 18684-24-7, C10H16N2O4, Mr 228.25, 250mg. Dipeptide-6. According to Shigemura et al. Pro-Hyp, a food-derived collagen peptide in human blood, stimulates the growth of fibroblasts from mouse skin. Nakatani et al. observed a chondroprotective effect of the dipeptide in mouse articular cartilage. Inoue et al. showed in a clinical study that ingestion of the dipeptides Pro-Hyp and Hyp-Gly could improve facial skin moisture and elasticity and reduce facial ageing.
Catalog No. 50-382-217 Supplier Bachem Supplier No. 4001630.0250
Add to Cart
Add to Cart
H-Pro-Hyp-OH, 18684-24-7, C10H16N2O4, Mr 228.25, 250mg. Dipeptide-6. According to Shigemura et al. Pro-Hyp, a food-derived collagen peptide in human blood, stimulates the growth of fibroblasts from mouse skin. Nakatani et al. observed a chondroprotective effect of the dipeptide in mouse articular cartilage. Inoue et al. showed in a clinical study that ingestion of the dipeptides Pro-Hyp and Hyp-Gly could improve facial skin moisture and elasticity and reduce facial ageing.