Learn More
CPC Scientific H-Pro-Leu-Gly-NHOH • HCl (hydrochloride salt) 250MG
SDP
Supplier:  CPC Scientific MMPS030B
Encompass
Affinity ligand for the purification of human collagenases. Inhibitor of MMP-8, Ki 19 M (Grams et al., 1995).SEQUENCE: H-Pro-Leu-Gly-NHOH HCl (hydrochloride salt)ONE-LETTER SEQUENCE: PLG-NHOH.HCLMOLECULAR FORMULA: C13H24N4O4HCl1MOLECULAR WEIGHT: 336.82STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [120928-08-7]SYNONYMS: RESEARCH AREA: CancerREFERENCES:W.M.Moore and C.A.Spilburg, Biochemistry, 25, 5189 (1986)F.Grams et al., Eur. J. Biochem., 228, 830 (1995)SKU(s): MMPS-030A, MMPS-030B
Catalog No. 50-210-9812