Learn More
Bachem H-Pro-Phe-Arg-AMC
SDP
Catalog No. 50260300
Encompass_Preferred
H-Pro-Phe-Arg-AMC Acetate, 65147-21-9, C30H37N7O5, Mr 575.67, 50mg. A highly sensitive, fluorogenic substrate for plasma as well as pancreatic and urinary kallikreins. PFR-AMC is also cleaved rapidly by soybean trypsin-like enzyme. Stock solutions of this substrate are best prepared in DMSO.
Catalog No. 50-260-300 Supplier Bachem Supplier No. 4004023.0050
Add to Cart
Add to Cart
H-Pro-Phe-Arg-AMC Acetate, 65147-21-9, C30H37N7O5, Mr 575.67, 50mg. A highly sensitive, fluorogenic substrate for plasma as well as pancreatic and urinary kallikreins. PFR-AMC is also cleaved rapidly by soybean trypsin-like enzyme. Stock solutions of this substrate are best prepared in DMSO.