Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC H-Pro-Val-OH | 52899-09-9 | 99.3% | 214.26 | 250 MG
SDP

Catalog No. 5000398520
Encompass_Preferred

H-Pro-Val-OH is a deprotonation dipeptide containing proline, capable of catalyzing the Michael addition reaction of acetone to trans-β-nitrostyrene. It can also function as a substrate for fibroblast enzymes and prolinase, with potential uses in biochemical analysis.

  • Purity: 99.3%
  • Molecular weight: 214.26
  • Formula: C10H18N2O3
  • Appearance: Solid
  • Color: Off-white to light brown
  • Sequence: Pro-Val
  • Solubility in H2O: ≥ 25 mg/mL (116.68 mM)
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Powder storage: -80°C for 2 years; -20°C for 1 year
  • In solvent storage: -80°C for 6 months; -20°C for 1 month (sealed storage, away from moisture)

Catalog No. 50-003-98520 Supplier Medchemexpress LLC Supplier No. 250MGHYP4566
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

H-Pro-Val-OH is a deprotonation dipeptide containing proline, capable of catalyzing the Michael addition reaction of acetone to trans-β-nitrostyrene. It can also function as a substrate for fibroblast enzymes and prolinase, with potential uses in biochemical analysis.

  • Purity: 99.3%
  • Molecular weight: 214.26
  • Formula: C10H18N2O3
  • Appearance: Solid
  • Color: Off-white to light brown
  • Sequence: Pro-Val
  • Solubility in H2O: ≥ 25 mg/mL (116.68 mM)
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Powder storage: -80°C for 2 years; -20°C for 1 year
  • In solvent storage: -80°C for 6 months; -20°C for 1 month (sealed storage, away from moisture)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.