Learn More
Medchemexpress LLC H-Pro-Val-OH | 52899-09-9 | 99.3% | 214.26 | 50 MG
SDP
Catalog No. 5000396696
Encompass_Preferred
H-Pro-Val-OH is a deprotonation dipeptide containing proline. It can catalyze the Michael addition reaction of acetone to trans-β-nitrostyrene. This compound can also act as a substrate for fibroblast enzymes and prolinase, offering potential applications in biochemical analysis.
  • Catalyzes Michael addition reactions
  • Acts as a substrate for fibroblast enzymes
  • Acts as a substrate for prolinase
  • Useful in biochemical analysis
Catalog No. 50-003-96696 Supplier Medchemexpress LLC Supplier No. HYP456650MG
H-Pro-Val-OH is a deprotonation dipeptide containing proline. It can catalyze the Michael addition reaction of acetone to trans-β-nitrostyrene. This compound can also act as a substrate for fibroblast enzymes and prolinase, offering potential applications in biochemical analysis.
  • Catalyzes Michael addition reactions
  • Acts as a substrate for fibroblast enzymes
  • Acts as a substrate for prolinase
  • Useful in biochemical analysis