Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TECHLAB™ H. PYLORI CHEK™ Diluent

Catalog No. 21279144
Change view
Click to view available options
Clia Complexity:
High
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Clia Complexity
21-279-144 High
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-279-144 Supplier TECHLAB™ Supplier No. 5051DIVD
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The H. PYLORI CHEK™ test is an enzyme immunoassay for the qualitative detection of Helicobacter pylori specific antigen. It is intended for use with human fecal specimens to aid in the diagnosis of H. pylori infection and to demonstrate loss of H. pylori antigen following treatment.

  • The test can be used with unpreserved fecal specimens and fecal specimens preserved in transport media from patients suspected of H. pylori infection.
  • Testing of patients to demonstrate loss of H. pylori antigen following treatment should be performed no sooner than 4 weeks after completion of the treatment regimen.
  • Test results should be taken into consideration by the physician in conjunction with the patient history and symptoms.

Specifications

CE Marker No
Clia Complexity High
Format 96-well Plate
Sample Type Fecal Sample
Sensitivity 100%
Type Diluent
Form Liquid Ready-to-Use
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.