Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H. PYLORI CHEK™ Stool Antigen ELISA Test

Catalog No. 23900556
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-900-556 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-900-556 Supplier Techlab Inc Supplier No. T5051
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The H. PYLORI CHEK™ Stool Antigen ELISA Test detects H. pylori specific antigen in human stool and provides excellent performance in determining active infection before and after eradication therapy. Developed and Manufactured by TECHLAB, Inc.

  • Sample types include fresh, frozen, Cary Blair, or C&S samples and can be stored refrigerated or at room temperature for 96 hours.
  • Provides excellent performance in determining active infection before and after eradication therapy
  • In compliance with current ACG guidelines
  • Detects current disease even in populations with high seropositivity

Specifications

CPT Code 87338
Clia Complexity High
Detectable Analytes H. pylori Antigen
Format Kit
Sample Type Fresh, Frozen, Transport Media (Cary Blair, C&S)
Includes 12 x 8-well strips with breakaway wells, Sample diluent, Control, Conjugate, Substrate, Wash Buffer
Quantity 96 Tests
Test Time 60 min
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.