Learn More
Drg International Inc DRG Helicobacter Pylori IgM ELISA RUO, 96 Well Kit
Supplier:  Drg International Inc EIA2111R
Encompass_Preferred
An enzyme immunoassay for the detection of IgM antibodies to Helicobacter pylori in serum. Purified H. pylori antigen is coated on the surface of microwells. Diluted serum is added to the wells, and the H. pylori IgM specific antibody, if present, binds to the antigen.
Catalog No. 50-194-7547