Learn More
Seracare Life Sciences Inc H PYLORI POS CONTROL
Supplier:  Seracare Life Sciences Inc 53700012
Encompass_Preferred
Heat-killed Helicobacter pylori cells (ATCC strain 43504). It consists of 1 mL of approximately 1% w/v wet packed cells in dextran solution. It is intended for use as a positive control in immunoassays designed for Helicobacter detection.
Catalog No. 50-674-64