Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H. PYLORI QUIK CHEK™ Stool Antigen Test

Catalog No. 23900555
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-900-555 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-900-555 Supplier Techlab Inc Supplier No. 30925
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

H. PYLORI QUIK CHEK™ Stool Antigen Test test detects H. pylori antigens in human stool and provides accurate results within 30 minutes. Developed and Manufactured by TECHLAB, Inc.

  • Can be used before treatment to diagnose and monitor infections and after treatment to confirm eradication
  • Offers high performance and flexible sample transportation conditions
  • Convenient – Results in 30 minutes without instrumentation using the standardized QUIK CHEK™ format
  • Flexible – Can be used with various sample types, including fresh, frozen, Cary Blair, or C&S samples and samples stored refrigerated or at room temperature for 96 hours
  • Reliable – Offers high sensitivity and specificity in determining active infection before and after eradication therapy

Specifications

CPT Code 87338
Clia Complexity Moderate
Detectable Analytes H. pylori Antigen
Format Kit
Sample Type Fresh, Frozen, Transport Media (Cary Blair, C & S)
Sensitivity 97%
Includes Membrane Devices, Conjugate, Diluent, Positive Control, Substrate, Wash Buffer, Transfer Pipettes, Applicator Sticks
Quantity 25 Tests
Specificity 1
Test Time 30 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.