Learn More
Diagnostic Biosystems H PYLORI RAPID STAIN H. Pylori Rapid Stain
Catalog No. 502407410
Encompass_Preferred
The H. Pylori Rapid Stain Kit is designed for demonstrating Helicobacter Pylori infected tissue. Kit may be used on formalin fixed, paraffin-embedded tissue.
Catalog No. 50-240-7410 Supplier Diagnostic Biosystems Supplier No. KT019
The H. Pylori Rapid Stain Kit is designed for demonstrating Helicobacter Pylori infected tissue. Kit may be used on formalin fixed, paraffin-embedded tissue.