Learn More
JANT PHARMACAL CORPORATION ACCUTEST H. Pylori Rapid Test Device (Whole Blood/Serum) *CLIA-Waived* 20 tests/kit
SDP
Catalog No. NC3456974
Encompass_Preferred
The Accutest H. pylori Whole Blood/Serum Cassette Test is a rapid lateral flow, qualitative immunoassay. It is intended for use at point of care facilities to detect the presence of IgG antibodies specific to Helicobacter pylori (H. pylori) in human blood or serum. It provides an aid in the diagnosis of infection by H. pylori.
This item is not returnable. View return policy
Catalog No. NC3456974 Supplier JANT PHARMACAL CORPORATION Supplier No. ID392
The Accutest H. pylori Whole Blood/Serum Cassette Test is a rapid lateral flow, qualitative immunoassay. It is intended for use at point of care facilities to detect the presence of IgG antibodies specific to Helicobacter pylori (H. pylori) in human blood or serum. It provides an aid in the diagnosis of infection by H. pylori.