Learn More
Biogenex Laboratories H. Pylori : ULC3R : 6mL
SDP
Supplier:  Biogenex Laboratories AM8805M
Encompass_Preferred
H. pylori are associated with B-type gastritis and some duodenal ulcers. Studies conducted by various researchers have shown that immunostaining for H. pylori with monoclonal antibodies is more reliable and easier to read than traditional H&E staining and significantly more sensitive than the Warthin-Starry silver stain, which is very complicated to perform. This antibody stains the bacilli in the lumen of an infected stomach in formalin-fixed, paraffin-embedded tissue sections.
Catalog No. 50-197-8773