Learn More
Bellco Glass, Inc. High Speed Vertical Lift Impeller-36L Complete
SDP
Catalog No. 501946200
Encompass_Preferred
H/S Vertical Lift Impeller-36L Complete. CONSIST OF; 7764-75036 & 7764-76036. DIMENSIONS: (SHAFT); 5/8"OD X 23 1/4" LONG, (DISK); 5 1/8"OD. 316L SST. MADE IN THE USA
Catalog No. 50-194-6200 Supplier Bellco Glass, Inc. Supplier No. 776476036
H/S Vertical Lift Impeller-36L Complete. CONSIST OF; 7764-75036 & 7764-76036. DIMENSIONS: (SHAFT); 5/8"OD X 23 1/4" LONG, (DISK); 5 1/8"OD. 316L SST. MADE IN THE USA