Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H-Ser But -OH, Advanced ChemTech

Catalog No. 01339499
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-339-499 1 g
01-339-497 5 g
01-339-498 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-339-499 Supplier Advanced Chem Tech Supplier No. IS74771G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18822-58-7
Molecular Formula C7H15NO3
Molecular Weight (g/mol) 161.20
InChI Key DDCPKNYKNWXULB-UHFFFAOYNA-N
IUPAC Name 2-amino-3-(tert-butoxy)propanoic acid
SMILES CC(C)(C)OCC(N)C(O)=O

Specifications

Color White to Off-White
Quantity 1 g
Grade Biochemical
Chemical Name or Material H-Ser But -OH
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.