Learn More
Medchemexpress LLC HY-126488 50mg Medchemexpress, H-Ser-His-OH CAS:67726-09-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12648850MG
Encompass_Preferred
Medchemexpress, HY-126488 50mg H-Ser-His-OH CAS:67726-09-4 H-Ser-His-OH is a short peptide with hydrolysis cleavage activity, an endogenous metabolite. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1108