Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC H-Ser-His-OH | 67726-09-4 | 99.8% | 1 ML
SDP

Catalog No. 5000341560
Encompass_Preferred

H-Ser-His-OH is a short peptide characterized by its hydrolysis cleavage activity and its role as an endogenous metabolite. This product offers high purity for reliable research applications.

  • High purity of 99.8%
  • Molecular weight of 242.23
  • Chemical formula C9H14N4O4
  • Functions as an inhibitor
  • Derived from endogenous metabolic pathways
  • Useful for metabolic disease research

Catalog No. 50-003-41560 Supplier Medchemexpress LLC Supplier No. HY12648810MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

H-Ser-His-OH is a short peptide characterized by its hydrolysis cleavage activity and its role as an endogenous metabolite. This product offers high purity for reliable research applications.

  • High purity of 99.8%
  • Molecular weight of 242.23
  • Chemical formula C9H14N4O4
  • Functions as an inhibitor
  • Derived from endogenous metabolic pathways
  • Useful for metabolic disease research

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.