Learn More
Medchemexpress LLC H-Ser-Phe-OH | 16875-28-8 | 99.8% | 252.27 | 10 MG
SDP
Catalog No. 5000397675
Encompass_Preferred
H-Ser-Phe-OH is a polypeptide used in peptide screening. This research tool identifies active peptides primarily through immunoassay techniques. It is applicable for protein interaction studies, functional analysis, and epitope screening, especially in agent research and development.
  • Polypeptide for peptide screening
  • Utilized in immunoassay techniques
  • Supports protein interaction studies
  • Enables functional analysis
  • Suitable for epitope screening
  • Valuable for agent research and development
Catalog No. 50-003-97675 Supplier Medchemexpress LLC Supplier No. HYP450910MG
H-Ser-Phe-OH is a polypeptide used in peptide screening. This research tool identifies active peptides primarily through immunoassay techniques. It is applicable for protein interaction studies, functional analysis, and epitope screening, especially in agent research and development.
  • Polypeptide for peptide screening
  • Utilized in immunoassay techniques
  • Supports protein interaction studies
  • Enables functional analysis
  • Suitable for epitope screening
  • Valuable for agent research and development