Learn More
Bachem H-Ser-Tyr-OH
SDP
Supplier:  Bachem 4003606.1000
Encompass_Preferred
H-Ser-Tyr-OH, 21435-27-8, C12H16N2O5, Mr 268.27, 1g. Ser-Tyr stimulated the uptake of deltorphin in SK-N-SH cells. The dipeptide forms a complex with Cu(II) acting as a tridentate ligand.
Catalog No. 50-259-384