Learn More
Apexbio Technology LLC H-Thr-Ome(Oil) HCl 39994-75-7 25g
SDP
Catalog No. 5000097135
Encompass_Preferred
H-Thr-Ome Oil HCl (CAS 39994-75-7) is a methyl ester hydrochloride derivative of the amino acid threonine Commonly utilized as a synthetic intermediate it facilitates peptide construction and structural modifications modulating peptide conformational properties and physiochemical interactions Researchers frequently use H-Thr-Ome Oil HCl to investigate structure-activity relationships (SAR) and elucidate peptide interactions with biomolecules Based on these chemical properties H-Thr-Ome Oil HCl holds research potential in peptide structure modification peptide-based compound synthesis and mechanism-oriented biomedical research
Catalog No. 50-000-97135 Supplier Apexbio Technology LLC Supplier No. A673925000
H-Thr-Ome Oil HCl (CAS 39994-75-7) is a methyl ester hydrochloride derivative of the amino acid threonine Commonly utilized as a synthetic intermediate it facilitates peptide construction and structural modifications modulating peptide conformational properties and physiochemical interactions Researchers frequently use H-Thr-Ome Oil HCl to investigate structure-activity relationships (SAR) and elucidate peptide interactions with biomolecules Based on these chemical properties H-Thr-Ome Oil HCl holds research potential in peptide structure modification peptide-based compound synthesis and mechanism-oriented biomedical research