Learn More
Medchemexpress LLC H-Thr(tBu)-OH | 4378-13-6 | MFCD00037766 | 97.0% | 175.23 g/mol | C8H17NO3 | 25 G
SDP
Catalog No. 5000304200
Encompass_Preferred
H-Thr(tBu)-OH is a t-butyl-protected threonine derivative used as an amino acid building block for peptide synthesis and small-molecule research. The protected hydroxyl group enables selective coupling and reduces side reactions during assembly; supplied as a white to off-white solid for laboratory use.
  • T-butyl protection on the hydroxyl group for selective peptide coupling.
  • High purity suitable for synthetic and analytical applications.
  • Solid form for convenient handling and storage.
  • Compatible with standard peptide coupling chemistries.
  • Useful as a building block in peptide and small-molecule synthesis.
Catalog No. 50-003-04200 Supplier Medchemexpress LLC Supplier No. 25GHYW016731
H-Thr(tBu)-OH is a t-butyl-protected threonine derivative used as an amino acid building block for peptide synthesis and small-molecule research. The protected hydroxyl group enables selective coupling and reduces side reactions during assembly; supplied as a white to off-white solid for laboratory use.
  • T-butyl protection on the hydroxyl group for selective peptide coupling.
  • High purity suitable for synthetic and analytical applications.
  • Solid form for convenient handling and storage.
  • Compatible with standard peptide coupling chemistries.
  • Useful as a building block in peptide and small-molecule synthesis.