Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC H-Trp-NH2.HCl | 5022-65-1 | 99.9% | 239.70 | 25 G
SDP

Catalog No. 5000453351
Encompass_Preferred

H-Trp-NH2.HCl is an endogenous metabolite. This product is for research use only and not sold to patients.

  • High purity (99.95%)
  • Molecular weight of 239.70
  • Chemical formula: C11H14ClN3O
  • Appearance: off-white to gray solid
  • Soluble in DMSO and H2O
  • Recommended storage at 4°C, sealed, away from moisture
  • In solvent, store at -80°C (6 months) or -20°C (1 month)

Catalog No. 50-004-53351 Supplier Medchemexpress LLC Supplier No. 25GHYW008766
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

H-Trp-NH2.HCl is an endogenous metabolite. This product is for research use only and not sold to patients.

  • High purity (99.95%)
  • Molecular weight of 239.70
  • Chemical formula: C11H14ClN3O
  • Appearance: off-white to gray solid
  • Soluble in DMSO and H2O
  • Recommended storage at 4°C, sealed, away from moisture
  • In solvent, store at -80°C (6 months) or -20°C (1 month)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.