Learn More
Medchemexpress LLC H-Trp-Phe-OH | 6686-02-8 | 99.9% | 351.40 | 50 MG
SDP
Catalog No. 5000398836
Encompass_Preferred
H-Trp-Phe-OH is a dipeptide consisting of tryptophan and phenylalanine, with antihypertensive properties. It inhibits angiotensin-converting enzyme (ACE), which increases nitric oxide (NO) levels and decreases endothelin-1 (ET-1) levels. It has also been shown to increase ovarian weight in female mice.
  • Consists of tryptophan and phenylalanine
  • Possesses antihypertensive properties
  • Inhibits angiotensin-converting enzyme (ACE)
  • Increases nitric oxide (NO) levels
  • Decreases endothelin-1 (ET-1) levels
  • Increases ovarian weight in female mice
Catalog No. 50-003-98836 Supplier Medchemexpress LLC Supplier No. HYP464150MG
H-Trp-Phe-OH is a dipeptide consisting of tryptophan and phenylalanine, with antihypertensive properties. It inhibits angiotensin-converting enzyme (ACE), which increases nitric oxide (NO) levels and decreases endothelin-1 (ET-1) levels. It has also been shown to increase ovarian weight in female mice.
  • Consists of tryptophan and phenylalanine
  • Possesses antihypertensive properties
  • Inhibits angiotensin-converting enzyme (ACE)
  • Increases nitric oxide (NO) levels
  • Decreases endothelin-1 (ET-1) levels
  • Increases ovarian weight in female mice