Learn More
Bachem H-Trp-Trp-Trp-OH
SDP
Supplier:  Bachem 4005236.0050
Encompass_Preferred
H-Trp-Trp-Trp-OH, 59005-82-2, C33H32N6O4, Mr 576.66, 50mg. Tritryptophan, WWW, a highly hydrophobic tripeptide used as a model in phase distribution studies.
Catalog No. 50-260-135