Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC H-Trp-Trp-Trp-OH | 59005-82-2 | 99.6% | 576.64 | 25 MG
SDP

Catalog No. 5000398764
Encompass_Preferred

H-Trp-Trp-Trp-OH is a tripeptide consisting of tryptophan, and it exhibits antibacterial activity.

  • Purity of 99.64%
  • Molecular weight of 576.64
  • Solid appearance
  • White to off-white color
  • Sequence: H-Trp-Trp-Trp-OH
  • Exhibits antibacterial activity
  • Stable for up to 2 years at -80°C in powder form
  • Soluble in DMSO at 100 mg/mL

Catalog No. 50-003-98764 Supplier Medchemexpress LLC Supplier No. 25MGHYP4648
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

H-Trp-Trp-Trp-OH is a tripeptide consisting of tryptophan, and it exhibits antibacterial activity.

  • Purity of 99.64%
  • Molecular weight of 576.64
  • Solid appearance
  • White to off-white color
  • Sequence: H-Trp-Trp-Trp-OH
  • Exhibits antibacterial activity
  • Stable for up to 2 years at -80°C in powder form
  • Soluble in DMSO at 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.