Learn More
Bachem H-Tyr-Arg-Gly-Asp-Ser-OH
SDP
Supplier:  Bachem 4030618.0005
Encompass_Preferred
H-Tyr-Arg-Gly-Asp-Ser-OH Trifluoroacetate, 134282-68-1, C24H36N8O10, Mr 596.60, 5mg. Y-R-G-D-S. PEG acrylate-based hydrogels have been modified with the RGD peptide YRGDS to improve their biocompatibility. YRGDS can be radioiodinated.
Catalog No. 50-259-713