Learn More
Bachem H-Tyr-His-OH
SDP
Supplier:  Bachem 4001846.1000
Encompass_Preferred
H-Tyr-His-OH, 3788-44-1, C15H18N4O4, Mr 318.33, 1g. Both single-dose administration and repetitive oral application of the dipeptide YH significantly decreased blood pressure in spontaneously hypertensive rats.
Catalog No. 50-259-400