Learn More
Medchemexpress LLC H-Tyr-Tyr-OH 100mg | 1050-28-8 | 100 MG
SDP
Catalog No. 5000228491
Encompass_Preferred
H-Tyr-Tyr-OH (L-tyrosyl-L-tyrosine) is a dipeptide inhibitor of angiotensin I-converting enzyme (ACE) used in biochemical and hypertension research. It exhibits ACE inhibitory activity (reported IC50 0.028 mg/mL) and is supplied as a high-purity solid or as ready-to-use DMSO solutions suitable for assay preparation.
  • High purity: 99.02%.
  • Molecular formula C18H20N2O5; molecular weight 344.36 g/mol.
  • Available as 100 mg solid and as DMSO solutions for convenient reconstitution.
  • Suitable for ACE activity assays and antihypertensive mechanism studies.
Catalog No. 50-002-28491 Supplier Medchemexpress LLC Supplier No. HY114782100MG
H-Tyr-Tyr-OH (L-tyrosyl-L-tyrosine) is a dipeptide inhibitor of angiotensin I-converting enzyme (ACE) used in biochemical and hypertension research. It exhibits ACE inhibitory activity (reported IC50 0.028 mg/mL) and is supplied as a high-purity solid or as ready-to-use DMSO solutions suitable for assay preparation.
  • High purity: 99.02%.
  • Molecular formula C18H20N2O5; molecular weight 344.36 g/mol.
  • Available as 100 mg solid and as DMSO solutions for convenient reconstitution.
  • Suitable for ACE activity assays and antihypertensive mechanism studies.