Learn More
Medchemexpress LLC H-Val-Ala-OH (Standard) | 27493-61-4 | 188.23 | 1G
SDP
Catalog No. 5000460289
Encompass_Preferred
H-Val-Ala-OH (Valyl-alanine) is a dipeptide formed from L-Valine and L-Alanine residues, functioning as a metabolite with a bitter taste. It is intended for research use only as an analytical standard for qualitative, quantitative, and methodological research experiments.
  • Dipeptide derived from L-Valine and L-Alanine
  • Acts as a metabolite
  • Possesses a bitter taste
  • Used as an analytical reference standard
  • Applicable in HPLC, GC, and MS research techniques
Catalog No. 50-004-60289 Supplier Medchemexpress LLC Supplier No. HYW0070351G
H-Val-Ala-OH (Valyl-alanine) is a dipeptide formed from L-Valine and L-Alanine residues, functioning as a metabolite with a bitter taste. It is intended for research use only as an analytical standard for qualitative, quantitative, and methodological research experiments.
  • Dipeptide derived from L-Valine and L-Alanine
  • Acts as a metabolite
  • Possesses a bitter taste
  • Used as an analytical reference standard
  • Applicable in HPLC, GC, and MS research techniques