Learn More
Medchemexpress LLC H-Val-Ala-OH (Standard) | 27493-61-4 | 188.23 | 50 G
SDP
Catalog No. 5000460376
Encompass_Preferred
H-Val-Ala-OH is a dipeptide formed from L-Valine and L-Alanine residues, functioning as a metabolite with a bitter taste. It can self-assemble into various ordered structures like square plates and wires in different solvents, with its self-assembly influenced by solvent type, concentration, and intermolecular hydrogen bonds.
  • Formed from L-Valine and L-Alanine residues
  • Functions as a metabolite
  • Possesses a bitter taste
  • Can self-assemble into various ordered structures
  • Self-assembly is influenced by solvent type, concentration, and intermolecular hydrogen bonds
  • Appears as a white to off-white solid
  • For research use only
Catalog No. 50-004-60376 Supplier Medchemexpress LLC Supplier No. HYW00703550G
H-Val-Ala-OH is a dipeptide formed from L-Valine and L-Alanine residues, functioning as a metabolite with a bitter taste. It can self-assemble into various ordered structures like square plates and wires in different solvents, with its self-assembly influenced by solvent type, concentration, and intermolecular hydrogen bonds.
  • Formed from L-Valine and L-Alanine residues
  • Functions as a metabolite
  • Possesses a bitter taste
  • Can self-assemble into various ordered structures
  • Self-assembly is influenced by solvent type, concentration, and intermolecular hydrogen bonds
  • Appears as a white to off-white solid
  • For research use only