Learn More
Medchemexpress LLC HY-114161A 10mg Medchemexpress, H-Val-Pro-Pro-OH (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY114161A10MG
Encompass_Preferred
Medchemexpress, HY-114161A 10mg H-Val-Pro-Pro-OH (TFA) CAS: H-Val-Pro-Pro-OH (TFA), a milk-derived proline peptides derivative, is an inhibitor of Angiotensin I converting enzyme (ACE), with an IC50 of 9 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9294