Learn More
Medchemexpress LLC H-Val-Pro-Pro-OH (TFA) | 2828433-08-3 | 98.0% | 425.40 | 50 MG
SDP
Catalog No. 5000436855
Encompass_Preferred
H-Val-Pro-Pro-OH (TFA) is a milk-derived proline peptide derivative that acts as an inhibitor of Angiotensin I converting enzyme (ACE), with an IC50 of 9 μM. For research use only, not sold to patients. H-Val-Pro-Pro-OH (TFA) can enhance insulin sensitivity and prevent insulin resistance in 3T3-F442A pre-adipocytes. It also possesses anti-hypertensive and anti-inflammatory functions, and can enhance the expression of glucose transporter 4 (GLUT4) in adipocytes and restore glucose uptake in TNF-treated adipocytes.
  • Inhibitor of Angiotensin I converting enzyme (ACE)
  • Enhances insulin sensitivity
  • Prevents insulin resistance in 3T3-F442A pre-adipocytes
  • Anti-hypertensive properties
  • Anti-inflammatory functions
  • Enhances expression of glucose transporter 4 (GLUT4) in adipocytes
  • Restores glucose uptake in TNF-treated adipocytes
Catalog No. 50-004-36855 Supplier Medchemexpress LLC Supplier No. HY114161A50MG
H-Val-Pro-Pro-OH (TFA) is a milk-derived proline peptide derivative that acts as an inhibitor of Angiotensin I converting enzyme (ACE), with an IC50 of 9 μM. For research use only, not sold to patients. H-Val-Pro-Pro-OH (TFA) can enhance insulin sensitivity and prevent insulin resistance in 3T3-F442A pre-adipocytes. It also possesses anti-hypertensive and anti-inflammatory functions, and can enhance the expression of glucose transporter 4 (GLUT4) in adipocytes and restore glucose uptake in TNF-treated adipocytes.
  • Inhibitor of Angiotensin I converting enzyme (ACE)
  • Enhances insulin sensitivity
  • Prevents insulin resistance in 3T3-F442A pre-adipocytes
  • Anti-hypertensive properties
  • Anti-inflammatory functions
  • Enhances expression of glucose transporter 4 (GLUT4) in adipocytes
  • Restores glucose uptake in TNF-treated adipocytes