Learn More
Bachem H-Val-Tyr-OH
SDP
Catalog No. 50259413
Encompass_Preferred
H-Val-Tyr-OH, 3061-91-4, C14H20N2O4, Mr 280.32, 1g. Antihypertensive dipeptide. Acts as ACE inhibitor, IC50 0.02 mg/ml. Val-Tyr showed an antihypertensive effect when administered orally to spontaneously hypertensive rats for 28 days.
Catalog No. 50-259-413 Supplier Bachem Supplier No. 4001976.0001
Add to Cart
Add to Cart
H-Val-Tyr-OH, 3061-91-4, C14H20N2O4, Mr 280.32, 1g. Antihypertensive dipeptide. Acts as ACE inhibitor, IC50 0.02 mg/ml. Val-Tyr showed an antihypertensive effect when administered orally to spontaneously hypertensive rats for 28 days.