Learn More
Across International Ai 100°C 7L Capacity Compact Heated Recirculator 110V
SDP
Catalog No. NC2012582
Encompass_Preferred
The Ai H100 series heated recirculators offer outstanding performance and high reliability as well as quiet operation and ease-of-use. Powerful force/suction pump provides constant flow rates, while the 2nd generation PID controller gives you the ideal solution when you need precise temperature control.
Catalog No. NC2012582 Supplier Across International Supplier No. H1007UL.110
The Ai H100 series heated recirculators offer outstanding performance and high reliability as well as quiet operation and ease-of-use. Powerful force/suction pump provides constant flow rates, while the 2nd generation PID controller gives you the ideal solution when you need precise temperature control.