Learn More
Sino Biological Influenza A H10N3 Hemagglutinin / HA Antibody, Rabbit PAb
Supplier:  Sino Biological 11693RP0120
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant H10N3 (A/duck/Hong Kong/786/1979) HA / Hemagglutinin (Catalog#11693-V08H; BAF46762.1; Met1-Ile525). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-2076