Learn More
Sino Biological Influenza A H10N8 Hemagglutinin / HA Neutralizing Antibody
Supplier:  Sino Biological 40359MH001200
Encompass_Preferred
It is a chimeric monoclonal antibody combining the constant domains of the human IgG1 molecule with mouse variable regions. The variable region was obtained from a mouse immunized with purified, recombinant Influenza A H10N8 (A/Jiangxi-Donghu/346/2013) Hemagglutinin / HA. The antibody was produced using recombinant antibody technology.
Catalog No. 50-241-1133