Learn More
Sino Biological Recombinant Human TP73/p73 Protein (Full Length, GST Tag)
Catalog No. 502573911
Encompass_Preferred
Recombinant full length human p73 gamma was expressed by baculovirus in Sf9 cells using an N-terminal GST tag.
Catalog No. 50-257-3911 Supplier Sino Biological Supplier No. P0830CG100
Recombinant full length human p73 gamma was expressed by baculovirus in Sf9 cells using an N-terminal GST tag.