Learn More
Sino Biological Influenza A H11N2 Hemagglutinin / HA Antibody, Rabbit PAb
Supplier:  Sino Biological 11705RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant H11N2 (A/duck/Yangzhou/906/2002) HA / Hemagglutinin (Catalog#11705-V08H; AAY85533.1; Met1-Lys528). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1644