Learn More
Sino Biological Influenza A H11N9 Hemagglutinin / HA Antibody, Rabbit PAb
Supplier:  Sino Biological 11704RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant H11N9 (A/mallard/Alberta/294/1977) HA / Hemagglutinin (Catalog#11704-V08H; ABB87228.1; Met 1-Lys 528). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-2099