Learn More
Sino Biological Influenza A H11N9 Hemagglutinin / HA Antibody, Rabbit PAb, Antigen Affinity Purified
Supplier:  Sino Biological 11704RP0220
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant H11N9 (A/mallard/Alberta/294/1977) HA / Hemagglutinin (Catalog#11704-V08H; ABB87228.1; Met 1-Lys 528). H11N9 (A/mallard/Alberta/294/1977) HA / Hemagglutinin specific IgG was purified by H11N9 (A/mallard/Alberta/294/1977) HA / Hemagglutinin affinity chromatography.
Catalog No. 50-241-0595