Learn More
Aviva Systems Biology Corp H13 Antibody - middle region (ARP49964_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49964_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against H13. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 1200006O09Rik, 4930443L17Rik, 5031424B04Rik, AV020344, H-13, Hm13, PSL3, SppApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-472