Learn More
Sino Biological Influenza A H13N8 Hemagglutinin / HA Antibody, Rabbit PAb
Supplier:  Sino Biological 11721RP0120
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant H13N8 (A/black-headed gull/Netherlands/1/00) HA / Hemagglutinin (Catalog#11721-V08H; AAV91212.1; Met 1-Lys 528). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8533